Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DPCD
Entrez Gene ID
423852
Gene Name
deleted in primary ciliary dyskinesia homolog (mouse)
Gene Aliases
-
Chromosome Location
Chr.6: 23757619 - 23765090 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277365.1 NP_001264294.1 4 - 5 453 110
NM_001277366.1 NP_001264295.1 4 - 5 435 110
GenBank mRNA BU353811.1 - 4 - 5 415 110
BU399154.1 - 4 - 5 422 110
BX931687.2 - 3 - 4 672 110
BX931759.1 - 4 - 5 430 110
BX932644.1 - 4 - 5 428 110
BX932782.1 - 4 - 5 445 110
BX933238.1 - 4 - 5 429 110
CR386779.1 - 4 - 5 411 110

View Full Product Details