Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CISD2
Entrez Gene ID
422714
Gene Name
CDGSH iron sulfur domain 2
Gene Aliases
ZCD2
Chromosome Location
Chr.4: 60944999 - 60951398 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_420668.6 XP_420668.1 2 - 3 378 72
GenBank mRNA BX932115.1 - 2 - 3 369 72
BX932825.2 - 2 - 3 326 72
BX936123.2 - 2 - 3 383 72
CR406249.1 - 2 - 3 372 72

View Full Product Details