Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC16A6
Entrez Gene ID
417435
Gene Name
solute carrier family 16 member 6
Gene Aliases
-
Chromosome Location
Chr.18: 7796068 - 7805377 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_415687.5 XP_415687.2 1 - 2 225 101
GenBank mRNA BX933032.1 - 1 - 2 134 101
BX935693.2 - 1 - 2 133 101

View Full Product Details