Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PBK
Entrez Gene ID
422003
Gene Name
PDZ binding kinase
Gene Aliases
-
Chromosome Location
Chr.3: 105111777 - 105120100 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277999.1 NP_001264928.1 2 - 3 175 133
XM_015285101.2 XP_015140587.1 3 - 4 205 133
XM_015285102.2 XP_015140588.1 3 - 4 183 133
GenBank mRNA BX930018.1 - 2 - 3 187 133
BX934024.1 - 2 - 3 175 133

View Full Product Details