Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PBK
Entrez Gene ID
422003
Gene Name
PDZ binding kinase
Gene Aliases
-
Chromosome Location
Chr.3: 105111777 - 105120100 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277999.1 NP_001264928.1 4 - 5 479 64
XM_015285101.2 XP_015140587.1 5 - 6 509 64
XM_015285102.2 XP_015140588.1 5 - 6 487 64
GenBank mRNA BU406153.1 - 1 - 2 54 64
BX930018.1 - 4 - 5 491 64
BX934024.1 - 4 - 5 479 64
CV860416.1 - - 114 64

View Full Product Details