Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM248
Entrez Gene ID
417549
Gene Name
transmembrane protein 248
Gene Aliases
C19H7ORF42, C7orf42
Chromosome Location
Chr.19: 5211136 - 5218594 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BX950729.1 - 2 - 3 374 97

View Full Product Details