Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DHRS7C
Entrez Gene ID
417304
Gene Name
dehydrogenase/reductase (SDR family) member 7C
Gene Aliases
-
Chromosome Location
Chr.18: 118988 - 128673 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277995.1 NP_001264924.1 4 - 5 585 111
GenBank mRNA BX929314.1 - 4 - 5 586 111
BX933187.2 - 1 - 2 230 111
BX935860.2 - 4 - 5 601 111

View Full Product Details