Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DHRS7C
Entrez Gene ID
417304
Gene Name
dehydrogenase/reductase (SDR family) member 7C
Gene Aliases
-
Chromosome Location
Chr.18: 118988 - 128673 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277995.1 NP_001264924.1 5 - 6 666 125
GenBank mRNA BX929314.1 - 5 - 6 667 125
BX935860.2 - 5 - 6 682 125

View Full Product Details