Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LPAR4
Entrez Gene ID
422149
Gene Name
lysophosphatidic acid receptor 4
Gene Aliases
GPR23
Chromosome Location
Chr.4: 1395220 - 1409368 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278049.1 NP_001264978.1 1 - 2 321 72
GenBank mRNA BX932026.2 - 1 - 2 164 72
CR353562.1 - 1 - 2 146 72

View Full Product Details