Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RRAD
Entrez Gene ID
415790
Gene Name
RRAD, Ras related glycolysis inhibitor and calcium channel regulator
Gene Aliases
ZGC:63471
Chromosome Location
Chr.11: 11211183 - 11216791 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277606.2 NP_001264535.2 2 - 3 398 99
GenBank mRNA BU307973.1 - 2 - 3 399 99
BX934073.1 - 2 - 3 399 99
BX950258.2 - 2 - 3 400 99

View Full Product Details