Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRKRIP1
Entrez Gene ID
417508
Gene Name
PRKR interacting protein 1 (IL11 inducible)
Gene Aliases
-
Chromosome Location
Chr.19: 4277201 - 4285736 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006214.2 NP_001006214.1 4 - 5 402 125
GenBank mRNA AJ719994.1 - 4 - 5 402 125
CR388669.1 - 4 - 5 370 125

View Full Product Details