The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199418.3 | NP_001186347.3 | 7 - 8 | 841 | 74 |
GenBank mRNA | CR523356.1 | - | 6 - 7 | 733 | 74 |