Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GABRQ
Entrez Gene ID
396288
Gene Name
gamma-aminobutyric acid (GABA) A receptor, theta
Gene Aliases
GABRB1, GABRB4
Chromosome Location
Chr.4: 10644261 - 10711020 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167703.1 NP_001161175.1 7 - 8 1050 124
NM_001167704.1 NP_001161176.1 7 - 8 1050 124
GenBank mRNA X56647.1 - - 1050 124
X56648.1 - - 753 124

View Full Product Details