Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COMMD8
Entrez Gene ID
422769
Gene Name
COMM domain containing 8
Gene Aliases
-
Chromosome Location
Chr.4: 66813955 - 66817743 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277821.1 NP_001264750.1 2 - 3 270 150
GenBank mRNA BU127447.1 - 2 - 3 247 150
BX929471.1 - 1 - 2 96 150
BX932299.2 - 2 - 3 213 150

View Full Product Details