Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DHRSX
Entrez Gene ID
428007
Gene Name
dehydrogenase/reductase X-linked
Gene Aliases
-
Chromosome Location
Chr.1: 129980980 - 130145451 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015277734.2 XP_015133220.1 3 - 4 393 101
GenBank mRNA BX931965.2 - 3 - 4 310 101
BX934631.1 - 3 - 4 288 101
BX950546.2 - 3 - 4 699 101
CR405833.1 - 2 - 3 178 101

View Full Product Details