Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PUSL1
Entrez Gene ID
419419
Gene Name
pseudouridylate synthase-like 1
Gene Aliases
-
Chromosome Location
Chr.21: 2351861 - 2382648 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277883.1 NP_001264812.1 7 - 8 866 85
NM_001277884.1 NP_001264813.1 7 - 8 917 85
XM_025142514.1 XP_024998282.1 7 - 8 851 85
XM_025142515.1 XP_024998283.1 7 - 8 1264 85
GenBank mRNA BX929622.1 - 7 - 8 917 85
BX931895.1 - 7 - 8 866 85

View Full Product Details