Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPG21
Entrez Gene ID
415526
Gene Name
spastic paraplegia 21 (autosomal recessive, Mast syndrome)
Gene Aliases
-
Chromosome Location
Chr.10: 18157255 - 18166713 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198603.1 NP_001185532.1 6 - 7 658 66
NM_001198604.1 NP_001185533.1 6 - 7 665 66
XM_015292090.2 XP_015147576.1 7 - 8 840 66
XM_025153856.1 XP_025009624.1 7 - 8 807 66
GenBank mRNA BX933431.2 - 7 - 8 832 66
BX934025.1 - 6 - 7 658 66
BX934064.1 - 6 - 7 665 66
BX935310.2 - 1 - 2 114 66

View Full Product Details