Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHMP5
Entrez Gene ID
420968
Gene Name
charged multivesicular body protein 5
Gene Aliases
-
Chromosome Location
Chr.2: 85638182 - 85646740 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277399.1 NP_001264328.1 7 - 8 697 72
GenBank mRNA BX934448.1 - 7 - 8 663 72
CR353120.1 - 7 - 8 670 72

View Full Product Details