Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NTS
Entrez Gene ID
417883
Gene Name
neurotensin
Gene Aliases
NT
Chromosome Location
Chr.1: 42207151 - 42223589 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277360.1 NP_001264289.1 3 - 4 544 122
XM_025148143.1 XP_025003911.1 4 - 5 460 122
XM_025148145.1 XP_025003913.1 4 - 5 553 122
GenBank mRNA BX931642.2 - 3 - 4 553 122
BX935235.1 - 3 - 4 492 122

View Full Product Details