Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPPP3
Entrez Gene ID
415669
Gene Name
tubulin polymerization promoting protein family member 3
Gene Aliases
-
Chromosome Location
Chr.11: 1221961 - 1224452 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015279191.2 XP_015134677.1 2 - 3 377 76
GenBank mRNA BX929488.1 - 1 - 2 196 76
BX933510.2 - 1 - 2 291 76
CR385779.1 - 1 - 2 303 76

View Full Product Details