Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VTG3
Entrez Gene ID
424534
Gene Name
vitellogenin 3
Gene Aliases
VYG3, VYGIII, vitellogenin-3, vtgIII
Chromosome Location
Chr.8: 16918916 - 16940019 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015290869.1 XP_015146355.1 8 - 9 1088 70
GenBank mRNA BX934727.2 - 8 - 9 967 70

View Full Product Details