Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXOSC3
Entrez Gene ID
423626
Gene Name
exosome component 3
Gene Aliases
-
Chromosome Location
Chr.6: 5127362 - 5131035 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199516.1 NP_001186445.1 3 - 4 624 113
GenBank mRNA BX259049.3 - 2 - 3 312 113
BX935636.2 - 3 - 4 584 113

View Full Product Details