Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRRG1
Entrez Gene ID
418582
Gene Name
proline rich and Gla domain 1
Gene Aliases
-
Chromosome Location
Chr.1: 114579211 - 114609343 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277736.1 NP_001264665.1 3 - 4 309 135
XM_015301827.2 XP_015157313.1 4 - 5 398 135
XM_015301833.2 XP_015157319.1 3 - 4 343 135
XM_015301839.2 XP_015157325.1 4 - 5 369 135
XM_015301845.2 XP_015157331.1 3 - 4 1418 135
XM_015301850.1 XP_015157336.1 3 - 4 237 135
XM_015301857.2 XP_015157343.1 3 - 4 1480 135
XM_015301861.1 XP_015157347.1 3 - 4 2827 135
GenBank mRNA BX935305.1 - 3 - 4 309 135
BX936218.2 - 3 - 4 309 135

View Full Product Details