The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_004939011.2 | XP_004939068.1 | 3 - 4 | 285 | 85 |
XM_417247.5 | XP_417247.3 | 2 - 3 | 141 | 85 | |
GenBank mRNA | CR386134.1 | - | 2 - 3 | 142 | 85 |
CR388921.1 | - | 2 - 3 | 130 | 85 | |
CR389175.1 | - | 2 - 3 | 129 | 85 |