Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC173
Entrez Gene ID
424161
Gene Name
coiled-coil domain containing 173
Gene Aliases
C7H2ORF77
Chromosome Location
Chr.7: 18603176 - 18612726 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA CR387407.1 - 6 - 7 980 138

View Full Product Details