Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LUM
Entrez Gene ID
417891
Gene Name
lumican
Gene Aliases
-
Chromosome Location
Chr.1: 44018283 - 44030483 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276357.1 NP_001263286.1 1 - 2 340 134
GenBank mRNA M80584.1 - 1 - 2 122 134

View Full Product Details