Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZHX3
Entrez Gene ID
419176
Gene Name
zinc fingers and homeoboxes 3
Gene Aliases
-
Chromosome Location
Chr.20: 4931004 - 4966608 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015296754.2 XP_015152240.1 1 - 2 221 134
GenBank mRNA BX930181.1 - 1 - 2 217 134
BX930230.1 - 1 - 2 143 134

View Full Product Details