Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MALL
Entrez Gene ID
421222
Gene Name
mal, T cell differentiation protein like
Gene Aliases
-
Chromosome Location
Chr.3: 3068307 - 3071184 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199428.1 NP_001186357.1 2 - 3 607 72
GenBank mRNA BX929438.2 - 2 - 3 608 72
BX930937.1 - 2 - 3 537 72

View Full Product Details