Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REEP3
Entrez Gene ID
423654
Gene Name
receptor accessory protein 3
Gene Aliases
-
Chromosome Location
Chr.6: 8461689 - 8499207 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199532.1 NP_001186461.1 1 - 2 91 117
GenBank mRNA BX932673.1 - 1 - 2 91 117

View Full Product Details