Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REEP3
Entrez Gene ID
423654
Gene Name
receptor accessory protein 3
Gene Aliases
-
Chromosome Location
Chr.6: 8461689 - 8499207 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199532.1 NP_001186461.1 5 - 6 483 152
XM_015288099.1 XP_015143585.1 5 - 6 461 152
XM_025151515.1 XP_025007283.1 4 - 5 393 152
GenBank mRNA BX932673.1 - 5 - 6 483 152
CR387172.1 - 5 - 6 647 152

View Full Product Details