Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REEP3
Entrez Gene ID
423654
Gene Name
receptor accessory protein 3
Gene Aliases
-
Chromosome Location
Chr.6: 8461689 - 8499207 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199532.1 NP_001186461.1 6 - 7 629 122
XM_015288099.1 XP_015143585.1 6 - 7 607 122
XM_025151515.1 XP_025007283.1 5 - 6 539 122
GenBank mRNA BU421613.1 - 1 - 2 122
BX932673.1 - 6 - 7 629 122

View Full Product Details