Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPIA4
Entrez Gene ID
423433
Gene Name
serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 4
Gene Aliases
SERPINA4
Chromosome Location
Chr.5: 45726288 - 45736369 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277492.1 NP_001264421.1 3 - 4 777 75
GenBank mRNA BX935008.1 - 3 - 4 771 75
KU180444.1 - 1 - 2 648 75

View Full Product Details