Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMP10
Entrez Gene ID
418981
Gene Name
matrix metallopeptidase 10
Gene Aliases
-
Chromosome Location
Chr.1: 183766311 - 183772932 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278089.1 NP_001265018.1 2 - 3 397 66
GenBank mRNA BX950347.1 - 2 - 3 387 66
CD218444.1 - 2 - 3 396 66

View Full Product Details