Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC170
Entrez Gene ID
421639
Gene Name
coiled-coil domain containing 170
Gene Aliases
C3H6orf97, C6ORF97, bA282P11.1
Chromosome Location
Chr.3: 49010559 - 49053032 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004935629.3 XP_004935686.1 3 - 4 326 82
XM_025149276.1 XP_025005044.1 3 - 4 626 82
XM_419678.6 XP_419678.4 3 - 4 625 82
GenBank mRNA BX931430.2 - 3 - 4 130 82

View Full Product Details