Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNGR1
Entrez Gene ID
418015
Gene Name
synaptogyrin 1
Gene Aliases
-
Chromosome Location
Chr.1: 50674047 - 50687764 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252278.3 NP_001239207.3 3 - 4 571 69
GenBank mRNA BX931567.2 - 3 - 4 559 69

View Full Product Details