Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYVE1
Entrez Gene ID
423038
Gene Name
lymphatic vessel endothelial hyaluronan receptor 1
Gene Aliases
XLKD1
Chromosome Location
Chr.5: 9149552 - 9160157 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199587.1 NP_001186516.1 1 - 2 251 91
GenBank mRNA BX932088.2 - 1 - 2 177 91
DT658132.1 - 1 - 2 281 91

View Full Product Details