Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEC13
Entrez Gene ID
416119
Gene Name
SEC13 homolog, nuclear pore and COPII coat complex component
Gene Aliases
SEC13L1
Chromosome Location
Chr.12: 19853537 - 19860212 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277841.1 NP_001264770.1 2 - 3 93 65
XM_015293127.2 XP_015148613.1 2 - 3 265 65
GenBank mRNA BX934554.2 - 2 - 3 93 65

View Full Product Details