Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYOM3
Entrez Gene ID
419690
Gene Name
myomesin 3
Gene Aliases
IFNLR1, IL28RA
Chromosome Location
Chr.23: 5779176 - 5826654 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_025143142.1 XP_024998910.1 1 - 2 235 99
GenBank mRNA BX935722.2 - 2 - 3 396 99

View Full Product Details