Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYOM3
Entrez Gene ID
419690
Gene Name
myomesin 3
Gene Aliases
IFNLR1, IL28RA
Chromosome Location
Chr.23: 5779176 - 5826654 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015297915.2 XP_015153401.1 4 - 5 952 105
XM_025143141.1 XP_024998909.1 4 - 5 952 105
XM_025143142.1 XP_024998910.1 4 - 5 683 105
XM_025143143.1 XP_024998911.1 4 - 5 952 105
XM_025143144.1 XP_024998912.1 4 - 5 952 105
GenBank mRNA BX935722.2 - 5 - 6 844 105
FJ947118.1 - 4 - 5 508 105
FJ947119.1 - 4 - 5 508 105
KF680105.1 - 4 - 5 508 105

Gene Symbol
IFNLR1
Entrez Gene ID
419694
Gene Name
interferon lambda receptor 1
Gene Aliases
CRF2-12, IL-28RA, IL28RA
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA KF680104.1 - 4 - 5 508 105

View Full Product Details