Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBE2D1
Entrez Gene ID
423641
Gene Name
ubiquitin conjugating enzyme E2 D1
Gene Aliases
-
Chromosome Location
Chr.6: 5935568 - 5951307 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199526.1 NP_001186455.1 4 - 5 241 113
XM_015288010.2 XP_015143496.1 5 - 6 359 113
XM_025151511.1 XP_025007279.1 4 - 5 255 113
GenBank mRNA BU411926.1 - 4 - 5 225 113
CR354339.1 - 4 - 5 241 113

View Full Product Details