Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCDH18
Entrez Gene ID
422438
Gene Name
protocadherin 18
Gene Aliases
-
Chromosome Location
Chr.4: 28645061 - 28654943 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004940935.3 XP_004940992.1 1 - 1 963 74
XM_420404.6 XP_420404.3 1 - 1 961 74
GenBank mRNA AF098475.1 - 1 - 1 58 74

View Full Product Details