Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- EMSY
- Entrez Gene ID
-
419090
- Gene Name
- EMSY, BRCA2 interacting transcriptional repressor
- Gene Aliases
-
2210018M11RIK, C11ORF30, C1H11orf30
- Chromosome Location
-
Chr.1: 195122953 - 195162521 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BX934563.1 |
- |
3 - 4 |
469 |
141 |
View Full Product Details