Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMARCE1
Entrez Gene ID
420047
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily e, member 1
Gene Aliases
-
Chromosome Location
Chr.27: 7303676 - 7318218 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006335.2 NP_001006335.2 4 - 5 256 136
XM_015299606.2 XP_015155092.1 4 - 5 263 136
GenBank mRNA AJ719694.1 - 4 - 5 256 136

View Full Product Details