Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIZ
Entrez Gene ID
421237
Gene Name
kizuna centrosomal protein
Gene Aliases
C20orf19, NCRNA00153, PLK1S1
Chromosome Location
Chr.3: 3599008 - 3639741 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006394.1 NP_001006394.1 10 - 11 1637 66
XM_015283363.2 XP_015138849.1 11 - 12 1709 66
XM_015283364.2 XP_015138850.1 11 - 12 1706 66
XM_015283365.2 XP_015138851.1 10 - 11 1643 66
XM_015283367.2 XP_015138853.1 11 - 12 3662 66
XM_015283369.2 XP_015138855.1 11 - 12 4143 66
XM_015283370.2 XP_015138856.1 12 - 13 2469 66
XM_015283371.2 XP_015138857.1 11 - 12 2372 66
XM_015283373.2 XP_015138859.1 9 - 10 1389 66
XM_025148640.1 XP_025004408.1 12 - 13 2530 66
GenBank mRNA AJ720271.1 - 10 - 11 1637 66
BX934707.2 - 5 - 6 740 66

View Full Product Details