Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIZ
Entrez Gene ID
421237
Gene Name
kizuna centrosomal protein
Gene Aliases
C20orf19, NCRNA00153, PLK1S1
Chromosome Location
Chr.3: 3599008 - 3639741 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006394.1 NP_001006394.1 6 - 7 1107 119
XM_015283363.2 XP_015138849.1 6 - 7 1110 119
XM_015283364.2 XP_015138850.1 6 - 7 1110 119
XM_015283365.2 XP_015138851.1 6 - 7 1110 119
XM_015283366.2 XP_015138852.1 6 - 7 1110 119
XM_015283367.2 XP_015138853.1 6 - 7 3063 119
XM_015283369.2 XP_015138855.1 6 - 7 3544 119
XM_015283370.2 XP_015138856.1 7 - 8 1870 119
XM_015283371.2 XP_015138857.1 6 - 7 1773 119
XM_015283373.2 XP_015138859.1 4 - 5 790 119
XM_025148639.1 XP_025004407.1 6 - 7 1110 119
XM_025148640.1 XP_025004408.1 7 - 8 1931 119
GenBank mRNA AJ720271.1 - 6 - 7 1107 119

View Full Product Details