Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIZ
Entrez Gene ID
421237
Gene Name
kizuna centrosomal protein
Gene Aliases
C20orf19, NCRNA00153, PLK1S1
Chromosome Location
Chr.3: 3599008 - 3639741 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006394.1 NP_001006394.1 7 - 8 1336 76
XM_015283363.2 XP_015138849.1 7 - 8 1339 76
XM_015283364.2 XP_015138850.1 7 - 8 1339 76
XM_015283365.2 XP_015138851.1 7 - 8 1339 76
XM_015283366.2 XP_015138852.1 7 - 8 1339 76
XM_015283367.2 XP_015138853.1 7 - 8 3292 76
XM_015283369.2 XP_015138855.1 7 - 8 3773 76
XM_015283370.2 XP_015138856.1 8 - 9 2099 76
XM_015283371.2 XP_015138857.1 7 - 8 2002 76
XM_015283373.2 XP_015138859.1 5 - 6 1019 76
XM_025148639.1 XP_025004407.1 7 - 8 1339 76
XM_025148640.1 XP_025004408.1 8 - 9 2160 76
GenBank mRNA AJ720271.1 - 7 - 8 1336 76
BX934707.2 - 2 - 3 436 76
CR387324.1 - 2 - 3 296 76

View Full Product Details