Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MXD4
Entrez Gene ID
422890
Gene Name
MAX dimerization protein 4
Gene Aliases
-
Chromosome Location
Chr.4: 82843102 - 82880589 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006460.1 NP_001006460.1 1 - 2 210 68
GenBank mRNA AJ719815.1 - 1 - 2 210 68

View Full Product Details