Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RRAS
Entrez Gene ID
423062
Gene Name
related RAS viral (r-ras) oncogene homolog
Gene Aliases
RRAS2
Chromosome Location
Chr.5: 10585170 - 10619898 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006466.1 NP_001006466.1 5 - 6 744 91
XM_015286388.2 XP_015141874.1 5 - 6 460 91
XM_015286389.1 XP_015141875.1 5 - 6 454 91
XM_015286390.2 XP_015141876.1 5 - 6 435 91
GenBank mRNA AJ720666.1 - 5 - 6 744 91

View Full Product Details