Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TOLLIP
Entrez Gene ID
423099
Gene Name
toll interacting protein
Gene Aliases
-
Chromosome Location
Chr.5: 14859899 - 14886241 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006471.1 NP_001006471.1 2 - 3 305 75
XM_015286612.2 XP_015142098.1 2 - 3 309 75
GenBank mRNA AJ720279.1 - 2 - 3 305 75

View Full Product Details