Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITPKA
Entrez Gene ID
395694
Gene Name
inositol-trisphosphate 3-kinase A
Gene Aliases
IP3KA
Chromosome Location
Chr.5: 24890429 - 24930838 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204881.1 NP_990212.1 2 - 3 691 134
GenBank mRNA AJ012219.1 - 2 - 3 691 134
U41445.2 - 2 - 3 121 134

View Full Product Details